Learn More
ABclonal Technology FOSB Rabbit pAb
SDP
Supplier:  ABclonal Technology A14696500UL
Encompass_Preferred
The Fos gene family consists of 4 members FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-04654