Learn More
Medchemexpress LLC Fosbretabulin disodium | 168555-66-6 | 99.9% | 440.29 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY174491MG
Encompass_Preferred
Fosbretabulin disodium (CA 4DP) is a tubulin destabilizing agent. It is a Combretastatin A4 proagent that selectively targets endothelial cells, induces regression of nascent tumor neovessels, reduces tumor blood flow, and causes central tumor necrosis.
  • Tubulin destabilizing agent
  • Selectively targets endothelial cells
  • Induces regression of nascent tumor neovessels
  • Reduces tumor blood flow
  • Causes central tumor necrosis
  • For research use only
Catalog No. 50-003-86686