Learn More
TARGETMOL CHEMICALS INC FOSBRETABULIN DISODIUM 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T627225MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fosbretabulin Disodium (CA 4P) a water-soluble prodrug of Combretastatin A4 (CA4) is a microtubule-targeting agent that binds (beta)-tubulin (Kd 0.4 uM). Fosbretabulin Disodium(Combretastatin A4 disodium phosphate) inhibits the polymerization of tubulin (IC50 2.4 uM) and also disrupts tumor vasculature. purity: 99%
Catalog No. 50-257-9462