Learn More
Medchemexpress LLC Foscarnet (sodium) | 63585-09-1 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB131810MM/1ML
Encompass_Preferred
Foscarnet sodium (Trisodium phosphonoformate) is an inhibitor of viral DNA polymerase activity, leading to reversible suppression of viral replication. It is also an antiherpesvirus agent used in the treatment of cytomegalovirus retinitis.
  • Inhibits viral DNA polymerase activity
  • Leads to reversible suppression of viral replication
  • Acts as an antiherpesvirus agent
  • Used in cytomegalovirus retinitis treatment
  • Inhibits Influenza A Victoria
  • Inhibits Influenza B HK
  • Inhibits Avian myeloblastosis virus
  • Inhibits HSV-1
Catalog No. 50-004-12052