Learn More
Medchemexpress LLC Foscarnet trisodium hexahydrate | 34156-56-4 | 99.5% | 300.04 | 10 G
Supplier: Medchemexpress LLC HYW01325610G
Foscarnet trisodium hexahydrate is an inhibitor of viral DNA polymerase activity, which leads to reversible suppression of viral replication. It is an antiherpesvirus agent used in cytomegalovirus retinitis.
- Viral DNA polymerase activity inhibitor
- Leads to reversible suppression of viral replication
- Antiherpesvirus agent
- Used in cytomegalovirus retinitis