Learn More
Medchemexpress LLC Fosfomycin (sodium) | 26016-99-9 | MFCD00056853 | 98.0% | 182.02 g/mol | C3H5Na2O4P | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01642010MM/1ML
Encompass_Preferred
Fosfomycin (sodium) is a water-soluble, broad-spectrum antibiotic reagent for in vitro microbiology and biochemical research. It irreversibly inhibits an early step in bacterial cell wall synthesis and is supplied both as a ready-to-use aqueous solution (10 mM, 1 mL) and in solid forms. Molecular formula C3H5Na2O4P; molecular weight 182.02 g/mol; CAS 26016-99-9.
  • Ready-to-use 10 mM aqueous solution for convenient dosing.
  • Also available as solid for custom preparation and storage.
  • Broad-spectrum antibacterial activity through cell wall synthesis inhibition.
  • Water-soluble for easy dilution and handling in aqueous assays.
  • Characterized reagent with accompanying datasheet and safety data sheet.
Catalog No. 50-003-02241