Learn More
Medchemexpress LLC Fosfomycin sodium | 26016-99-9 | MFCD00056853 | >98.0% | 182.02 g/mol | C3H5Na2O4P | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0164205G
Encompass_Preferred
Fosfomycin sodium is the disodium salt of fosfomycin supplied as an analytical/reference standard for laboratory and microbiological use. It appears as a white to off-white powder, is water-soluble, and is commonly reported at about 98.0% purity for use in assays, method development, and susceptibility testing.
  • High-purity reference standard suitable for analytical applications.
  • Broad-spectrum antibiotic activity useful for microbiology controls.
  • Water-soluble sodium salt form for easy dissolution.
  • Consistent lot-to-lot performance for assay reproducibility.
  • Available in multiple pack sizes to support small-scale and bulk needs.
Catalog No. 50-003-02201