Learn More
TARGETMOL CHEMICALS INC FOSPHENYTOIN 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T61354100MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Fosphenytoin is a phosphate prodrug that can be used as an alternative to intravenous phenytoin for the acute intervention of seizures.Fosphenytoin is hydrolyzed to anticonvulsant phenytoin when administered parenterally.Fosphenytoin can be used in a variety of delivery modes such as intravenous and intraperitoneal.Fosphenytoin can be used in a variety of delivery modes such as intravenous and intraperitoneal. purity: 100%
Catalog No. 50-257-9527