Learn More
TARGETMOL CHEMICALS INC FOSTAMATINIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T611510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fostamatinib (R788)(IC50 of 41 nM) which is a prodrug of the active metabolite R406 is a Syk inhibitor. It does not work on Lyn. purity: 98%
Catalog No. 50-257-6598