Learn More
Medchemexpress LLC Fostamatinib | 901119-35-5 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY13038A1MG
Encompass_Preferred
Fostamatinib (R788) is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. This compound also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
  • Oral proagent of the active compound R406
  • Orally available and competitive Syk/FLT3 inhibitor
  • Inhibits Lyn and Lck
Catalog No. 50-003-40644