Learn More
Medchemexpress LLC Fostamatinib | 901119-35-5 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13038A10MM/1ML
Encompass_Preferred
Fostamatinib (R788) is an orally available proagent of R406, which is a competitive inhibitor of Syk and FLT3. It also demonstrates inhibitory activity against Lyn and Lck.
  • Syk/FLT3 inhibitor
  • Also inhibits Lyn and Lck
  • Active compound R406
  • High purity of 99.7%
  • Pre-prepared 10 mM solution in DMSO
  • Stable under recommended storage conditions
  • Used in various clinical trials
Catalog No. 50-003-39853