Learn More
Medchemexpress LLC Fostamatinib disodium hexahydrate | 1025687-58-4 | MFCD15146370 | 98.9% | C23H24FN6Na2O9P | 10MG
SDP
Supplier:  Medchemexpress LLC HY1303810MG
Encompass_Preferred
Fostamatinib disodium hexahydrate is an analytical/reference standard of the oral prodrug fostamatinib supplied for research and analytical use. The compound corresponds to CAS 1025687-58-4 and is the hexahydrate disodium salt of the prodrug whose active metabolite R406 is a competitive Syk/FLT3 inhibitor. The Certificate of Analysis reports purity 98.9% and molecular formula C23H24FN6Na2O9P.
  • Analytical/reference standard for research and method development.
  • High purity (98.9%) for reliable analytical and QC results.
  • Prodrug of R406; active metabolite exhibits Syk and FLT3 inhibition.
  • Supplied as a hexahydrate salt with COA documentation for traceability.
  • Store and handle according to COA recommendations to maintain stability.
Catalog No. 50-002-60509