Learn More
Medchemexpress LLC Fostamatinib disodium hexahydrate | 914295-16-2 | 99.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13038B50MG
Encompass_Preferred
Fostamatinib disodium hexahydrate (R788) is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM.
  • Oral proagent of active compound R406
  • Syk/FLT3 inhibitor
  • High bioavailability and rapid absorption
  • Purity of 99.5%
  • Available as a white to gray solid
  • Provided with a data sheet, COA, and SDS
Catalog No. 50-003-40900