Learn More
Selleck Chemical LLC Fostamatinib R788-2mg
SDP
Supplier:  Selleck Chemical LLC S26252MG
Encompass_Preferred
Fostamatinib (R788), a prodrug of the active metabolite R406, is a Syk inhibitor with IC50 of 41 nM, strongly inhibits Syk but not Lyn, 5-fold less potent to Flt3. Phase 3.
Catalog No. 50-238-9967