Learn More
Medchemexpress LLC Fostamatinib (R788) | 901119-35-5 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13038A100MG
Encompass_Preferred
Fostamatinib (R788) is an oral proagent of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
  • Inhibits Syk and FLT3, as well as Lyn and Lck.
  • Highly bioavailable and rapidly absorbed, converting completely to R406 in plasma.
  • Intended for research purposes only; not for sale to patients.
Catalog No. 50-003-38607