Learn More
Medchemexpress LLC HY-15440B 5mg Medchemexpress, Fostemsavir Tris CAS:864953-39-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15440B5MG
Encompass_Preferred
Medchemexpress, HY-15440B 5mg Fostemsavir Tris CAS:864953-39-9 Fostemsavir Tris (BMS-663068 (Tris)) is the phosphonooxymethyl prodrug of BMS-626529. Fostemsavir Tris (BMS-663068 (Tris)) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4+ T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9939