Learn More
Penn Tool Inc FOWLER 6"/150MM ULTRA CAL IV
SDP
Supplier:  Penn Tool Inc 541004441
Encompass
electronic caliper, water resistant to ip54 spec, direct rs232c ouput, includes:6"/150mm, -accuracy:.0008"/20um, resolution:.0005"/.01mm, repeatability:.0005"/.01mm, protection rating iec 60529:ip54
Catalog No. NC1677319