Learn More
MSC Fowler Gage Block Set, 81 Pc, AS-1 Grade, 0.05 to 4 in Range, Steel
Supplier:  MSC 95971529
Encompass
This gage block set is designed for precision measurement applications. It features 81 steel pieces with an AS-1 accuracy grade, covering a measurement range from 0.05 to 4 inches. The blocks are square-shaped and conform to ASME B89.1.9-2002 standards. This set is individually serialized, includes certification, and comes with a fitted case for convenient storage and transport.
  • 81 pieces included for comprehensive measurement options.
  • Constructed from durable steel.
  • AS-1 Accuracy Grade ensures high precision.
  • Measurement range of 0.05 to 4 inches.
  • Square block shape.
  • Individually serialized blocks.
  • Includes a fitted case.
  • Adheres to ASME B89.1.9-2002 standards.
  • Certification included.
Catalog No. 50-005-30393