Learn More
Cayman Chemical FOXM1 InhIbItr III 25mg
SDP
Supplier:  Cayman Chemical 3693225 MG
Encompass_Preferred
An inhibitor of FOXM1 DNA-binding activity (IC50 = 22.5 µM); binds to FOXM1 and is selective for inhibiting FOXM1 transcriptional activity over FOXA1, FOXA2, FOXP2, and GATA-1 transcriptional activity; inhibits the growth of MCF-7, MDA-MB-231, and PEO1 cancer cells (GI50s = 18, 21.8, and 18.1 µM, respectively)
Catalog No. 50-291-4988