Learn More
Beckman Coulter Beckman Coulter FoxP3-PC7 Conjugated Antibody, Clone 259D, 0.5 mL, ASR
Supplier: Beckman Coulter B46032
Beckman Coulter FoxP3-PC7 is an analyte-specific reagent (ASR) monoclonal antibody intended for detection of the FOXP3 transcription factor by flow cytometry. It is supplied as a PC7-conjugated antibody (clone 259D) in a 0.5 mL vial for research use.
- Detects FOXP3 transcription factor in human cells.
- Conjugated to PC7 for flow cytometry detection.
- Clone 259D.
- Supplied as a 0.5 mL liquid reagent.
- Isotype: IgG1 (mouse).
- Provided as an analyte-specific reagent for research use.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.