Learn More
Bellco Glass, Inc. Cap Torque Tool, 3/89" Square Drive, 316 Stainless Steel, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800422
Encompass_Preferred
VersaCap Torque Tool – 3/8" Drive for 83B Cap, SS316L Double Bagged for Sterile EnvironmentsTorque tool is here to ensure that you get a consistent, leak-proof seal every time. It’s designed to work perfectly with our VersaCap Technology, applying the right amount of torque for secure and repetitive usage.The tool is available for multiple cap sizes, including 38-430, GL45, 53B, 83B, and 120mm. Each tool fits precisely and is easy to use, helping you apply the same torque every time.DesignedUSA-foxx-logoFeaturesPrevents under- or over-tightening during assembly, ensuring a secure fit.Made with passivated SS316L which prevents from aggressive cleaning agents and acids.Lightweight and easy to use.ApplicationsFilling lines and cleanroom operations require validated closure force.
Catalog No. 50-255-0340