Learn More
Medchemexpress LLC Fp-biotin | 259270-28-5 | MFCD02094222 | 99.8% | 592.75 g/mol | C27H50FN4O5PS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1369245MG
Encompass_Preferred
FP-Biotin is a biotin-labeled organophosphorus activity-based probe for detecting and quantifying serine hydrolase enzymes in biochemical and proteomic assays. Supplied as a solid or as a 10 mM solution in DMSO, it enables enrichment of biotinylated peptides by avidin/streptavidin capture for downstream analysis of FAAH, ABHD6, and MAG-lipase activity.
  • High purity (99.8%) suitable for analytical applications.
  • Biotin tag enables efficient avidin/streptavidin-based enrichment.
  • Detects and quantifies serine hydrolase activity such as FAAH, ABHD6, MAG-lipase.
  • Available as solid and ready-to-use DMSO solution formats.
  • Compatible with plasma and proteomic workflows for biomarker discovery.
Catalog No. 50-002-21957