Learn More
TARGETMOL CHEMICALS INC FPH2 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T229225MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FPH2 (BRD-9424) (BRD-9424) is a small molecule promoting differentiation of iPS-derived hepatocytes. purity: 99%
Catalog No. 50-258-0913