Learn More
Selleck Chemical LLC FPR Agonist 43 E7571-1MG
SDP
Supplier:  Selleck Chemical LLC E75711MG
Encompass_Preferred
FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist
Catalog No. 50-000-43440