Learn More
Selleck Chemical LLC FPS-ZM1 50mg 945714-67-0
SDP
Supplier:  Selleck Chemical LLC S818550MG
Encompass_Preferred
FPS-ZM1 is a blood-brain-barrier permeant, non-toxic, tertiary amide compound which is a high affinity RAGE-specific inhibitor, blocking A? binding to the V domain of RAGE. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5768