Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Multitube Frame, 150-mm Scale; Four Tube, Isolated Flow, 316 Stainless Steel
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0321552
Encompass
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require. These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.
Catalog No. 50-196-1070