Learn More
World Precision Instrument FrameWorks Polished Stainless Steel Post, 50cm
SDP
Supplier:  World Precision Instrument 503071
Encompass
These high quality components are made of stainless steel and polymer that resists organic solvents and corrosion. They can be easily assembled to make a stand-alone setup for student laboratory equipment or to make a complicated frame for research labs. Use these components to custom build your own stands for laboratory glassware, manipulator stands or any other laboratory application you can envision.
Catalog No. 50-252-1320