Learn More
Bioworld Fraser Broth Base, 500 g
Supplier:  Bioworld 306200271
Encompass_Preferred
Fraser Broth BaseFor the isolation and selective enrichment of Listeria species from food and environmental speciesDoes NOT contain Fraser Supplement SolutionpH: 7.2Working Concentration: 55 g/LDehydrated Appearance: tan, homogeneous powderPrepared Appearance: medium amber brothSoluble in boiling waterComposition:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Beef Extract Yeast Extract Sodium Chloride Disodium Phosphate Monopotassium Phosphate Esculin Acriflavine Nalidixic Acid Lithium Chloride
Catalog No. 50-254-305