Learn More
Medchemexpress LLC Frataxin/FXN Human 100ug
SDP
Supplier:  Medchemexpress LLC HYP72266100UG
Encompass_Preferred
Frataxin/FXN protein activates the core iron-sulfur cluster assembly complex and accelerates sulfur transfer for [2Fe-2S] cluster assembly It plays a key role in the de novo synthesis of clusters delivering persulfide to the ISCU Frataxin/FXN Protein Human (His Myc) is the recombinant human-derived Frataxin/FXN protein expressed by E coli with C-Myc N-10 His labeled tag
Catalog No. 50-001-82006