Learn More
ABclonal Technology Frataxin/FXN Rabbit mAb
SDP
Supplier:  ABclonal Technology A25507100UL
Encompass_Preferred
This nuclear gene encodes a mitochondrial protein which belongs to the FRATAXIN family The protein functions in regulating mitochondrial iron transport and respiration The expansion of intronic trinucleotide repeat GAA from 8-33 repeats to -gt 90 repeats results in Friedreich ataxia Alternative splicing results in multiple transcript variants
Catalog No. 50-000-13136