Learn More
Selleck Chemical LLC FRAX1036 5mg 1432908-05-8
SDP
Supplier:  Selleck Chemical LLC S79895MG
Encompass_Preferred
FRAX-1036 is a potent and selective PAK inhibitor with biochemical potency (Ki) of 23.3 nM and 72.4 nM against PAK1 and PAK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5514