Learn More
Selleck Chemical LLC FRAX486 5mg 1232030-35-1
SDP
Supplier:  Selleck Chemical LLC S78075MG
Encompass_Preferred
FRAX486 is a potent p21-activated kinase (PAK) inhibitor with IC50 values of 14, 33, 39 and 575 nM for PAK1, PAK2, PAK3 and PAK4 respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5276