Learn More
Apexbio Technology LLC FRAX597, 10mM (in 1mL DMSO). Cas: 1286739-19-2 MFCD: MFCD25976723. PAK inhibitor,potent and ATP-competitive
SDP
Supplier:  Apexbio Technology LLC B11625.1
Encompass_Preferred
FRAX597 is a small-molecule inhibitor of the group 1 p21-activated kinases (PAKs) with IC50 values of 8nM, 13nM and 19nM, respectively for PAK1, PAK2 and PAK3 [1]. The PAKs family includes two sub groups: 1 and 2. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5208