Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fraxidin | 525-21-3 | MFCD00017492 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN39075MG

Encompass_Preferred

Fraxidin is a naturally occurring coumarin isolated from the roots of Jatropha podagrica. It is recognized for its antibacterial activity, particularly against Bacillus subtilis, where it exhibits an inhibition zone of 12 mm at a concentration of 20 μg/disk. This compound has a molecular weight of 222.19 and a molecular formula of C11H10O5, typically presenting as an off-white to light yellow solid. It is intended for research use only and belongs to the Phenylpropanoid, Coumarin, Phenol, and Monophenol classifications.

  • Naturally occurring coumarin
  • Exhibits antibacterial activity against Bacillus subtilis
  • Identified by CAS 525-21-3
  • Off-white to light yellow solid appearance
  • Molecular formula: C11H10O5
  • Molecular weight: 222.19

Catalog No. 50-004-19295


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.