Learn More
Medchemexpress LLC HY-N0579 5mg Medchemexpress, Fraxin CAS:524-30-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05795MG
Encompass_Preferred
Medchemexpress, HY-N0579 5mg Fraxin CAS:524-30-1 Fraxin isolated from Acer tegmentosum, F. ornus or A. hippocastanum, is a glucoside of fraxetin and reported to exert potent anti-oxidative stress action, anti-inflammatory and antimetastatic properties. Fraxin shows its antioxidative effect through inhibition of cyclo AMP phosphodiesterase enzyme. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0290