Learn More
Medchemexpress LLC Fraxin (Fraxoside) | 524-30-1 | 370.31 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0579100MG
Encompass_Preferred
Fraxin, also known as Fraxoside, is isolated from Cortex Fraxini. It is a glucoside of fraxetin and has been reported to possess potent anti-oxidative stress, anti-inflammatory, and antimetastatic properties. Fraxin exhibits its antioxidative effect by inhibiting the cyclo AMP phosphodiesterase enzyme.
  • Isolated from Cortex Fraxini
  • Glucoside of fraxetin
  • Exerts potent anti-oxidative stress action
  • Possesses anti-inflammatory properties
  • Shows antimetastatic properties
  • Inhibits cyclo AMP phosphodiesterase enzyme
Catalog No. 50-003-88728