Learn More
Medchemexpress LLC Fraxin (Fraxoside) | 524-30-1 | MFCD00010093 | 370.31 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN057910MM/1ML
Encompass_Preferred
Fraxin, also known as Fraxoside, is isolated from Cortex Fraxini and is a glucoside of fraxetin. It is reported to possess potent anti-oxidative stress, anti-inflammatory, and antimetastatic properties through the inhibition of cyclo AMP phosphodiesterase enzyme.
  • Possesses anti-oxidative stress action
  • Exhibits anti-inflammatory properties
  • Shows antimetastatic properties
  • Inhibits cyclo AMP phosphodiesterase enzyme
Catalog No. 50-003-88729