Learn More
MOLECULAR DEPOT LLC Free Bilirubin - 1 mL
Supplier:  MOLECULAR DEPOT LLC K2010001B
Encompass_Preferred
Free Bilirubin for Interference Testing a highly concentrated solution of free bilirubin (20 mg/mL) provided with the corresponding diluent This solution can be spiked into specimens to test free bilirubin interference Instructions with sample calculations are also provided For in vitro research use only
Catalog No. 50-000-13339