Learn More
Cusabio Technology LLC free haemoglobin f-Hb
Supplier:  Cusabio Technology LLC CSBEQ028141MO
Encompass_Preferred
The Mouse free haemoglobin (f-Hb) ELISA Kit is designed to quantitatively measure f-Hb levels in serum and plasma samples using a competitive assay format This kit offers a detection range of 0 781 g/mL to 50 g/mL with a sensitivity of 0 177 g/mL ensuring precise measurement of low-abundance targets The procedure requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm providing reliable detection of f-Hb for research or diagnostic applications Its standardized protocol and consistent performance make it suitable for studying hemolysis-related conditions transfusion reactions or other scenarios requiring accurate haemoglobin quantification in biological fluids
Catalog No. 50-000-52149