Learn More
Cusabio Technology LLC Mouse Free Thyroxine,FT4 ELISA Kit
Supplier:  Cusabio Technology LLC CSBE05080M
Encompass_Preferred
This Mouse FT4 ELISA Kit was designed for the quantitative measurement of Mouse FT4 protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 4 pmol/L-64 pmol/L and the sensitivity is 1.08 pmol/L. One kit contains 96 wells.
Catalog No. NC2952806