Learn More
TWD Scientific LLC Freezer Box, Twdbiox, 2In, 81 Well, Polypropylene, Assorted Colors, Removable Lid, Numeric Grid On Box And Lid, 20/Case.
SDP
Supplier:  TWD Scientific LLC BX909081
Encompass_Preferred
2 Inch 81 Well Polypropylene Freezer Box (Dimensions: 132X132X53Mm) With Removable Lid. Boxes/Lids Have Numeric Grid For Identification Purposes. Boxes Come In Assorted Colors (Clear, Pink, Blue, Green And Yellow). 20/Case, Price/Case
Catalog No. 50-228-5096