Learn More
Cole-Parmer Argos Technologies PolarSafe Metal Freezer Box, Stainless Steel, 3"; 1/Ea
Supplier:  Cole-Parmer 0439809
Encompass_Preferred
These metal freezer boxes are guaranteed not to rust. They feature drain holes that is ideal for storing samples in most liquid nitrogen tanks offered on the market. They come in stainless steel or aluminum, and the boxes can be stored in the same rack configurations as the two and three inch cardboard boxes. Cardboard dividers can be added, too, depending on the application.
Catalog No. 50-196-0679