Learn More
Research Products International Corp Freezer Drawer Rack for 50 ml Tubes, 78 Tube Capacity, 22 x 5 1/2 x 10 Inches
SDP
Supplier:  Research Products International Corp 181515
Encompass
Stainless steel freezer racks feature easy slide out drawers to provide quick and convenient access to samples. Racks accommodate a large number of tubes for improved freezer capacity and eliminate the need for storing individual boxes. Racks fit standard upright freezers.
Catalog No. 50-476-494