Learn More
Alkali Scientific Alkali Scientific™ Freezer Drawer Racks for Storage of Bottles, 3 Drawers, Drawer Area Capacity: 15″ x 4 1/2″
SDP
Supplier:  Alkali Scientific UFDRB033
Encompass
For Storage Bottles The UFDR-B03-3 Upright Freezer Drawer Rack for storage bottles features 3 drawers and is constructed of corrosion resistant stainless steel. Drawer racks facilitate both access to and storage of bottles without having to remove the whole rack from the freezer. These freezer racks for storage bottles include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Dimensions L x H x W : 16 11/16 x 9 1/8 x 5 1/2 inch (424 x 231 x 140mm)Capacity: Drawer Area 15 x 4 1/2 (Per Drawer) 383 x 114mm
Catalog No. 50-302-8439