Learn More
Alkali Scientific Freezer Rack Locking Device
SDP
Supplier:  Alkali Scientific DR23
Encompass_Preferred
The Freezer Rack Locking Device is designed to securely hold storage racks in place within laboratory or industrial freezers. Ideal for preventing accidental displacement during handling, this locking mechanism enhances safety and organization in temperature sensitive environments. Key Features: Durable Construction: Made from high quality, cold resistant materials to withstand extreme low temperatures. Secure Fit: Ensures racks remain stable and locked in position during use or transport. Universal Compatibility: Fits most standard freezer storage racks and boxes. Tool Free Operation: Easy to install and remove without the need for additional tools. Corrosion Resistant Finish: Built to resist moisture and chemical exposure common in laboratory settings.
Catalog No. 50-006-62310