Learn More
Stellar Scientific Freezer rack with drawer for 15 mL centrifuge tubes. Holds 208 tubes 26 3/4 x 9 15/16 x 5 1/2 ( D x H x W) 1/EA
SDP
Supplier:  Stellar Scientific CIFR15208
Encompass_Preferred
PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market.These unusual racks are great for labs that store samples in 15 or 50ML tubes and need quick access. Each tube is inserted into its own slot allowing for fast and easy identification. No more digging though boxes that are locked frozen in ice. Many different configurations available to support just 15 or 50s or a combination of tubes. Holds 208 tubes 26 3/4 x 9 15/16 x 5 1/2 ( D x H x W)
Catalog No. NC1322289