Learn More
Research Products International Corp Freezer Rack for 100 Place Slides, Stainless Steel, 3 x 4 Array, Dimensions (Inches) 8 1/2 x 22 1/8 x 6 5/8H
SDP
Supplier:  Research Products International Corp 247528
Encompass
The slotted Microscope Slide Freezer Rack provides convenient storage for slide racks. Helps save time by keeping you organized in a cost efficient manner. The highest quality racks, constructed of corrosion resistant stainless steel, includes handles. Racks for 100 place slide boxes fit all 7 x 8 1/4 x 1 3/8 boxes.
Catalog No. 50-550-271