Learn More
Shockwatch Div Mri FreezeSafe 5°C
SDP
Supplier:  Shockwatch Div Mri E01FRE005C02P1
Encompass_Preferred
FreezeSafe temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably low temperature levels, allowing you to determine if product quality has been compromised. Temperature threshold: 5C/41F. Indicator turns from clear to magenta withing 120 minutes. Accuracy +/-1C. Shelf life 12 months. Single-use.
Catalog No. NC2598359