Learn More
Medchemexpress LLC Freselestat (ONO-6818) | 208848-19-5 | 99.7% | 452.5 g/mol | C23H28N6O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1565210MG
Encompass_Preferred
Freselestat is a potent neutrophil elastase inhibitor supplied as a white-to-off-white solid for research use. It is intended for biochemical and pharmacological studies of neutrophil elastase activity and related inflammatory pathways.
  • Potent neutrophil elastase inhibitor with low-nanomolar activity.
  • High purity appropriate for research applications.
  • Solid powder form, easy to weigh and dissolve for assays.
  • Available in small packaging sizes suitable for preclinical experiments.
Catalog No. 50-002-85780