Learn More
Bachem Mca-Ala-Pro-Lys(Dnp)-OH
SDP
Supplier:  Bachem 4042638.0001
Encompass_Preferred
Mca-Ala-Pro-Lys(Dnp)-OH, 305336-82-7, C32H36N6O12, Mr 696.67, 1mg. Mca-APK(Dnp). FRET substrate for angiotensin-converting enzyme 2 (ACE2). Mca-APK(Dnp)-OH is used for monitoring the activity of the enzyme in plasma, urine, heart, and lungs.
Catalog No. 50-194-6738